Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153487.3 | NP_705691.1 | 9 - 10 | 3078 | 102 |
XM_006715056.3 | XP_006715119.1 | 9 - 10 | 2022 | 102 | |
XM_017010734.1 | XP_016866223.1 | 9 - 10 | 2022 | 102 | |
GenBank mRNA | AF478693.1 | - | 9 - 10 | 3078 | 102 |