Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002581.4 | NP_002572.2 | 1 - 2 | 795 | 102 |
XM_017014784.1 | XP_016870273.1 | 1 - 2 | 835 | 102 | |
GenBank mRNA | BC078657.1 | - | 1 - 2 | 425 | 102 |
CB960120.1 | - | 1 - 2 | 452 | 102 | |
X68280.1 | - | 1 - 2 | 186 | 102 |