Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001174116.1 | NP_001167587.1 | 20 - 21 | 5181 | 77 |
NM_001174117.1 | NP_001167588.1 | 18 - 19 | 3273 | 77 | |
NM_015263.3 | NP_056078.2 | 20 - 21 | 5181 | 77 | |
XM_005254255.1 | XP_005254312.1 | 20 - 21 | 5244 | 77 | |
XM_005254256.1 | XP_005254313.1 | 20 - 21 | 5244 | 77 | |
XM_017022034.1 | XP_016877523.1 | 20 - 21 | 4917 | 77 | |
GenBank mRNA | AB020663.2 | - | 8 - 9 | 2558 | 77 |
AF389880.1 | - | 20 - 21 | 5163 | 77 | |
AK123284.1 | - | 3 - 4 | 1008 | 77 | |
BC140781.1 | - | 18 - 19 | 3138 | 77 | |
BC144539.1 | - | 20 - 21 | 5046 | 77 |