Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001174116.1 | NP_001167587.1 | 29 - 30 | 7616 | 147 |
NM_001174117.1 | NP_001167588.1 | 27 - 28 | 5705 | 147 | |
NM_015263.3 | NP_056078.2 | 29 - 30 | 7613 | 147 | |
XM_005254255.1 | XP_005254312.1 | 29 - 30 | 7679 | 147 | |
XM_005254256.1 | XP_005254313.1 | 29 - 30 | 7676 | 147 | |
XM_017022034.1 | XP_016877523.1 | 29 - 30 | 7352 | 147 | |
GenBank mRNA | AB020663.2 | - | 17 - 18 | 4993 | 147 |
AF389880.1 | - | 29 - 30 | 7595 | 147 | |
BC016032.1 | - | 6 - 7 | 1765 | 147 | |
BC140781.1 | - | 27 - 28 | 5570 | 147 | |
BC144539.1 | - | 29 - 30 | 7481 | 147 |