Assay Design



Gene Transcripts

Gene Symbol
DMXL2
Entrez Gene ID
23312
Gene Name
Dmx like 2
Gene Aliases
PEPNS, RC3
Chromosome Location
Chr.15: 51447724 - 51622833 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001174116.1 NP_001167587.1 6 - 7 791 85
NM_001174117.1 NP_001167588.1 6 - 7 791 85
NM_015263.3 NP_056078.2 6 - 7 791 85
XM_005254255.1 XP_005254312.1 6 - 7 854 85
XM_005254256.1 XP_005254313.1 6 - 7 854 85
XM_011521402.2 XP_011519704.1 6 - 7 854 85
XM_017022034.1 XP_016877523.1 6 - 7 527 85
GenBank mRNA AF389880.1 - 6 - 7 773 85
BC140781.1 - 6 - 7 656 85
BC144539.1 - 6 - 7 656 85

View Full Product Details