Assay Design



Gene Transcripts

Gene Symbol
TAS2R9
Entrez Gene ID
50835
Gene Name
taste 2 receptor member 9
Gene Aliases
T2R9, TRB6
Chromosome Location
Chr.12: 10809094 - 10810168 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023917.2 NP_076406.1 1 - 1 857 107
GenBank mRNA BC069066.1 - 1 - 1 764 107
BC095519.1 - 1 - 1 858 107

View Full Product Details