Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031966.3 | NP_114172.1 | 5 - 6 | 952 | 69 |
GenBank mRNA | AK289431.1 | - | 5 - 6 | 841 | 69 |
AM851053.1 | - | 5 - 6 | 699 | 69 | |
BC006510.2 | - | 5 - 6 | 794 | 69 | |
BT020128.1 | - | 5 - 6 | 699 | 69 | |
M25753.1 | - | 5 - 6 | 788 | 69 |