Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000899.4 | NP_000890.1 | 3 - 4 | 390 | 140 |
NM_003994.5 | NP_003985.2 | 3 - 4 | 390 | 140 | |
GenBank mRNA | AF119835.1 | - | 3 - 4 | 337 | 140 |
AF400436.1 | - | 3 - 4 | 378 | 140 | |
AF400437.1 | - | 3 - 4 | 337 | 140 | |
AK290319.1 | - | 3 - 4 | 386 | 140 | |
AK293002.1 | - | 3 - 4 | 306 | 140 | |
BC069733.1 | - | 3 - 4 | 351 | 140 | |
BC069783.1 | - | 3 - 4 | 352 | 140 | |
BC069797.1 | - | 3 - 4 | 347 | 140 | |
BC074725.2 | - | 3 - 4 | 235 | 140 | |
BC126166.1 | - | 3 - 4 | 345 | 140 | |
BC143899.1 | - | 3 - 4 | 345 | 140 | |
DC320486.1 | - | 3 - 4 | 390 | 140 | |
M59964.1 | - | 3 - 4 | 378 | 140 |