Assay Design



Gene Transcripts

Gene Symbol
U2SURP
Entrez Gene ID
23350
Gene Name
U2 snRNP associated SURP domain containing
Gene Aliases
SR140, fSAPa
Chromosome Location
Chr.3: 143001524 - 143060725 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080415.1 NP_001073884.1 21 - 22 2324 147
NM_001320219.1 NP_001307148.1 21 - 22 2321 147
NM_001320220.1 NP_001307149.1 20 - 21 2280 147
NM_001320222.1 NP_001307151.1 22 - 23 2389 147
XM_005247248.4 XP_005247305.1 21 - 22 2330 147
XM_017006037.1 XP_016861526.1 17 - 18 1932 147
XM_017006038.1 XP_016861527.1 20 - 21 2216 147
XM_017006039.1 XP_016861528.1 14 - 15 1659 147
GenBank mRNA AB002330.1 - 21 - 22 2223 147
AK057679.1 - 8 - 9 2318 147
AK074327.1 - 14 - 15 1588 147
BC006474.1 - 21 - 22 2220 147
BC111692.1 - 20 - 21 2651 147

View Full Product Details