Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206999.1 | NP_001193928.1 | 44 - 45 | 5682 | 67 |
NM_007174.2 | NP_009105.1 | 43 - 44 | 5556 | 67 | |
XM_006719206.2 | XP_006719269.1 | 44 - 45 | 5664 | 67 | |
XM_011537783.1 | XP_011536085.1 | 45 - 46 | 5754 | 67 | |
XM_011537784.1 | XP_011536086.1 | 45 - 46 | 5754 | 67 | |
XM_011537785.1 | XP_011536087.1 | 45 - 46 | 5709 | 67 | |
XM_011537787.1 | XP_011536089.1 | 43 - 44 | 5538 | 67 | |
XM_011537788.1 | XP_011536090.1 | 43 - 44 | 5538 | 67 | |
XM_011537789.1 | XP_011536091.1 | 37 - 38 | 4889 | 67 | |
XM_011537790.2 | XP_011536092.1 | 36 - 37 | 4457 | 67 | |
XM_011537791.2 | XP_011536093.1 | 36 - 37 | 5442 | 67 | |
XM_011537792.2 | XP_011536094.1 | 35 - 36 | 5497 | 67 | |
XM_017018735.1 | XP_016874224.1 | 44 - 45 | 5709 | 67 | |
XM_017018736.1 | XP_016874225.1 | 45 - 46 | 5709 | 67 | |
XM_017018737.1 | XP_016874226.1 | 44 - 45 | 5664 | 67 | |
XM_017018738.1 | XP_016874227.1 | 34 - 35 | 4998 | 67 | |
XM_017018739.1 | XP_016874228.1 | 34 - 35 | 5773 | 67 | |
GenBank mRNA | AB023166.2 | - | 35 - 36 | 5497 | 67 |
AY257469.1 | - | 43 - 44 | 5553 | 67 | |
AY681966.1 | - | 44 - 45 | 5682 | 67 |