Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000857.3 | NP_000848.1 | 2 - 3 | 249 | 132 |
NM_001291951.1 | NP_001278880.1 | 2 - 3 | 249 | 132 | |
NM_001291953.1 | NP_001278882.1 | 3 - 4 | 435 | 132 | |
NM_001291954.1 | NP_001278883.1 | 2 - 3 | 249 | 132 | |
XM_011531901.1 | XP_011530203.1 | 2 - 3 | 163 | 132 | |
XM_017008130.1 | XP_016863619.1 | 2 - 3 | 440 | 132 | |
XM_017008132.1 | XP_016863621.1 | 2 - 3 | 440 | 132 | |
GenBank mRNA | AF020340.1 | - | 2 - 3 | 132 | 132 |
AK300296.1 | - | 2 - 3 | 201 | 132 | |
AK307838.1 | - | 3 - 4 | 377 | 132 | |
BC047620.1 | - | 2 - 3 | 163 | 132 | |
DC317249.1 | - | 2 - 3 | 272 | 132 | |
DC342619.1 | - | 2 - 3 | 250 | 132 | |
X66533.1 | - | 2 - 3 | 169 | 132 |