Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002738.6 | NP_002729.2 | 6 - 7 | 890 | 85 |
NM_212535.2 | NP_997700.1 | 6 - 7 | 890 | 85 | |
GenBank mRNA | BC036472.1 | - | 6 - 7 | 850 | 85 |
M13975.1 | - | 6 - 7 | 757 | 85 | |
X06318.1 | - | 6 - 7 | 829 | 85 | |
X07109.1 | - | 6 - 7 | 829 | 85 |