Assay Design



Gene Transcripts

Gene Symbol
PRDM2
Entrez Gene ID
7799
Gene Name
PR domain 2
Gene Aliases
HUMHOXY1, KMT8, MTB-ZF, RIZ, RIZ1, RIZ2
Chromosome Location
Chr.1: 13698875 - 13825079 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012231.4 NP_036363.2 2 - 3 867 149
NM_015866.4 NP_056950.2 2 - 3 867 149
GenBank mRNA U17838.1 - 2 - 3 866 149

View Full Product Details