Assay Design



Gene Transcripts

Gene Symbol
FGD4
Entrez Gene ID
121512
Gene Name
FYVE, RhoGEF and PH domain containing 4
Gene Aliases
CMT4H, FRABP, ZFYVE6
Chromosome Location
Chr.12: 32399523 - 32646050 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304480.1 NP_001291409.1 15 - 16 2350 159
NM_001304481.1 NP_001291410.1 15 - 16 2316 159
NM_001304483.1 NP_001291412.1 16 - 17 2540 159
NM_001304484.1 NP_001291413.1 16 - 17 2559 159
NM_139241.3 NP_640334.2 15 - 16 2440 159
XM_005253304.4 XP_005253361.1 15 - 16 2576 159
XM_005253307.3 XP_005253364.1 14 - 15 2552 159
XM_005253308.4 XP_005253365.1 14 - 15 2604 159
XM_005253309.1 XP_005253366.1 14 - 15 2064 159
XM_005253310.3 XP_005253367.1 11 - 12 1234 159
XM_011520554.1 XP_011518856.1 15 - 16 2224 159
XM_011520555.1 XP_011518857.1 15 - 16 2266 159
XM_011520556.1 XP_011518858.1 16 - 17 2495 159
XM_011520557.1 XP_011518859.1 13 - 14 1658 159
XM_011520558.2 XP_011518860.1 12 - 13 1378 159
XM_011520559.2 XP_011518861.1 11 - 12 1220 159
XM_017018803.1 XP_016874292.1 15 - 16 2576 159
XM_017018804.1 XP_016874293.1 14 - 15 2404 159
XM_017018805.1 XP_016874294.1 13 - 14 1554 159
GenBank mRNA AK057294.1 - 15 - 16 2323 159
AK297025.1 - 15 - 16 2316 159
AK303334.1 - 15 - 16 2321 159
AL832064.1 - 11 - 12 1270 159
BC045552.1 - 16 - 17 2413 159

View Full Product Details