Assay Design



Gene Transcripts

Gene Symbol
FGD4
Entrez Gene ID
121512
Gene Name
FYVE, RhoGEF and PH domain containing 4
Gene Aliases
CMT4H, FRABP, ZFYVE6
Chromosome Location
Chr.12: 32399523 - 32646050 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304480.1 NP_001291409.1 8 - 9 1579 141
NM_001304481.1 NP_001291410.1 8 - 9 1545 141
NM_001304483.1 NP_001291412.1 9 - 10 1769 141
NM_001304484.1 NP_001291413.1 9 - 10 1788 141
NM_139241.3 NP_640334.2 8 - 9 1669 141
XM_005253304.4 XP_005253361.1 8 - 9 1805 141
XM_005253307.3 XP_005253364.1 7 - 8 1781 141
XM_005253308.4 XP_005253365.1 7 - 8 1833 141
XM_005253309.1 XP_005253366.1 7 - 8 1293 141
XM_005253310.3 XP_005253367.1 4 - 5 463 141
XM_011520554.1 XP_011518856.1 8 - 9 1453 141
XM_011520555.1 XP_011518857.1 8 - 9 1495 141
XM_011520556.1 XP_011518858.1 9 - 10 1724 141
XM_011520557.1 XP_011518859.1 6 - 7 887 141
XM_011520558.2 XP_011518860.1 5 - 6 607 141
XM_011520559.2 XP_011518861.1 4 - 5 449 141
XM_017018803.1 XP_016874292.1 8 - 9 1805 141
XM_017018804.1 XP_016874293.1 7 - 8 1633 141
XM_017018805.1 XP_016874294.1 6 - 7 783 141
GenBank mRNA AK057294.1 - 8 - 9 1552 141
AK297025.1 - 8 - 9 1545 141
AK303334.1 - 8 - 9 1550 141
AL832064.1 - 4 - 5 499 141
AY367054.1 - 6 - 7 717 141

View Full Product Details