Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001520.2 | NP_001001520.1 | 5 - 6 | 760 | 67 |
NM_032631.3 | NP_116020.1 | 5 - 6 | 760 | 67 | |
XM_011528375.2 | XP_011526677.1 | 5 - 6 | 1120 | 67 | |
XM_011528376.2 | XP_011526678.1 | 5 - 6 | 1120 | 67 | |
XM_011528377.2 | XP_011526679.1 | 5 - 6 | 1120 | 67 | |
GenBank mRNA | AB208892.1 | - | 3 - 4 | 459 | 67 |
AF294267.1 | - | 5 - 6 | 667 | 67 | |
AL520441.3 | - | 5 - 6 | 705 | 67 | |
AY358600.1 | - | 5 - 6 | 669 | 67 | |
BC000755.2 | - | 5 - 6 | 650 | 67 | |
BC009449.1 | - | 2 - 3 | 299 | 67 | |
BI905780.1 | - | 5 - 6 | 698 | 67 |