Assay Design



Gene Transcripts

Gene Symbol
FBN3
Entrez Gene ID
84467
Gene Name
fibrillin 3
Gene Aliases
-
Chromosome Location
Chr.19: 8065402 - 8149846 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321431.1 NP_001308360.1 29 - 30 3847 67
NM_032447.4 NP_115823.3 29 - 30 4122 67
XM_017027372.1 XP_016882861.1 29 - 30 3846 67
XM_017027373.1 XP_016882862.1 29 - 30 4105 67
XM_017027374.1 XP_016882863.1 28 - 29 4009 67
XM_017027375.1 XP_016882864.1 29 - 30 4105 67
XM_017027376.1 XP_016882865.1 29 - 30 4105 67
XM_017027377.1 XP_016882866.1 10 - 11 1172 67
XM_017027378.1 XP_016882867.1 2 - 3 399 67
XM_017027379.1 XP_016882868.1 30 - 31 4231 67
GenBank mRNA AB053450.2 - 28 - 29 3728 67
AY165863.1 - 29 - 30 4122 67
AY165864.1 - 29 - 30 3847 67
AY165865.1 - 28 - 29 3993 67
BC153882.1 - 28 - 29 3728 67
CN369190.1 - 1 - 2 66 67

View Full Product Details