Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321431.1 | NP_001308360.1 | 33 - 34 | 4354 | 95 |
NM_032447.4 | NP_115823.3 | 33 - 34 | 4629 | 95 | |
XM_017027372.1 | XP_016882861.1 | 33 - 34 | 4353 | 95 | |
XM_017027373.1 | XP_016882862.1 | 33 - 34 | 4612 | 95 | |
XM_017027374.1 | XP_016882863.1 | 32 - 33 | 4516 | 95 | |
XM_017027376.1 | XP_016882865.1 | 33 - 34 | 4612 | 95 | |
XM_017027377.1 | XP_016882866.1 | 14 - 15 | 1679 | 95 | |
XM_017027378.1 | XP_016882867.1 | 6 - 7 | 906 | 95 | |
XM_017027379.1 | XP_016882868.1 | 34 - 35 | 4738 | 95 | |
GenBank mRNA | AB053450.2 | - | 32 - 33 | 4235 | 95 |
AY165863.1 | - | 33 - 34 | 4629 | 95 | |
AY165864.1 | - | 33 - 34 | 4354 | 95 | |
AY165865.1 | - | 32 - 33 | 4500 | 95 | |
BC153882.1 | - | 32 - 33 | 4235 | 95 | |
CN369190.1 | - | 5 - 6 | 573 | 95 |