Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321431.1 | NP_001308360.1 | 35 - 36 | 4592 | 123 |
NM_032447.4 | NP_115823.3 | 35 - 36 | 4867 | 123 | |
XM_017027372.1 | XP_016882861.1 | 35 - 36 | 4591 | 123 | |
XM_017027373.1 | XP_016882862.1 | 35 - 36 | 4850 | 123 | |
XM_017027374.1 | XP_016882863.1 | 34 - 35 | 4754 | 123 | |
XM_017027375.1 | XP_016882864.1 | 34 - 35 | 4727 | 123 | |
XM_017027376.1 | XP_016882865.1 | 35 - 36 | 4850 | 123 | |
XM_017027377.1 | XP_016882866.1 | 16 - 17 | 1917 | 123 | |
XM_017027378.1 | XP_016882867.1 | 8 - 9 | 1144 | 123 | |
XM_017027379.1 | XP_016882868.1 | 36 - 37 | 4976 | 123 | |
GenBank mRNA | AB053450.2 | - | 34 - 35 | 4473 | 123 |
AY165863.1 | - | 35 - 36 | 4867 | 123 | |
AY165864.1 | - | 35 - 36 | 4592 | 123 | |
AY165865.1 | - | 34 - 35 | 4738 | 123 | |
BC153882.1 | - | 34 - 35 | 4473 | 123 |