Assay Design



Gene Transcripts

Gene Symbol
FBN3
Entrez Gene ID
84467
Gene Name
fibrillin 3
Gene Aliases
-
Chromosome Location
Chr.19: 8065402 - 8149846 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321431.1 NP_001308360.1 46 - 47 5921 106
NM_032447.4 NP_115823.3 46 - 47 6196 106
XM_017027372.1 XP_016882861.1 46 - 47 5920 106
XM_017027373.1 XP_016882862.1 46 - 47 6179 106
XM_017027374.1 XP_016882863.1 45 - 46 6083 106
XM_017027375.1 XP_016882864.1 45 - 46 6056 106
XM_017027377.1 XP_016882866.1 27 - 28 3246 106
XM_017027378.1 XP_016882867.1 19 - 20 2473 106
XM_017027379.1 XP_016882868.1 47 - 48 6305 106
GenBank mRNA AB053450.2 - 45 - 46 5802 106
AK096539.1 - 1 - 2 123 106
AK131282.1 - 4 - 5 504 106
AY165863.1 - 46 - 47 6196 106
AY165864.1 - 46 - 47 5921 106
AY165865.1 - 45 - 46 6067 106
BC153882.1 - 45 - 46 5802 106

View Full Product Details