Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282410.1 | NP_001269339.1 | 1 - 2 | 894 | 62 |
NM_002712.2 | NP_002703.1 | 1 - 2 | 894 | 62 | |
GenBank mRNA | BC000910.2 | - | 1 - 2 | 67 | 62 |
BC012397.1 | - | 1 - 2 | 60 | 62 | |
BC013001.2 | - | 1 - 2 | 72 | 62 | |
BT007296.1 | - | 1 - 2 | 56 | 62 | |
BT020134.1 | - | 1 - 2 | 56 | 62 | |
Z50749.1 | - | 1 - 2 | 71 | 62 |