Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000901.4 | NP_000892.2 | 4 - 5 | 2372 | 96 |
XM_011531975.1 | XP_011530277.1 | 4 - 5 | 2384 | 96 | |
XM_011531976.2 | XP_011530278.1 | 4 - 5 | 2681 | 96 | |
XM_011531977.2 | XP_011530279.1 | 4 - 5 | 2111 | 96 | |
XM_011531978.2 | XP_011530280.1 | 4 - 5 | 2384 | 96 | |
XM_017008217.1 | XP_016863706.1 | 4 - 5 | 2669 | 96 | |
GenBank mRNA | AB209056.1 | - | 3 - 4 | 1618 | 96 |
M16801.1 | - | 4 - 5 | 2225 | 96 |