Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000901.4 | NP_000892.2 | 6 - 7 | 2875 | 110 |
NM_001166104.1 | NP_001159576.1 | 5 - 6 | 2524 | 110 | |
XM_011531975.1 | XP_011530277.1 | 6 - 7 | 2887 | 110 | |
XM_011531976.2 | XP_011530278.1 | 6 - 7 | 3184 | 110 | |
XM_011531977.2 | XP_011530279.1 | 6 - 7 | 2614 | 110 | |
XM_017008217.1 | XP_016863706.1 | 6 - 7 | 3172 | 110 | |
GenBank mRNA | AB209056.1 | - | 5 - 6 | 2121 | 110 |
AJ315515.1 | - | 2 - 3 | 248 | 110 | |
BC111758.1 | - | 5 - 6 | 2228 | 110 | |
M16801.1 | - | 6 - 7 | 2728 | 110 |