Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000901.4 | NP_000892.2 | 7 - 8 | 3008 | 68 |
NM_001166104.1 | NP_001159576.1 | 6 - 7 | 2657 | 68 | |
XM_011531975.1 | XP_011530277.1 | 7 - 8 | 3020 | 68 | |
XM_011531976.2 | XP_011530278.1 | 7 - 8 | 3317 | 68 | |
XM_011531977.2 | XP_011530279.1 | 7 - 8 | 2747 | 68 | |
XM_017008217.1 | XP_016863706.1 | 7 - 8 | 3305 | 68 | |
GenBank mRNA | AB209056.1 | - | 6 - 7 | 2254 | 68 |
AJ315514.1 | - | 5 - 6 | 2363 | 68 | |
BC111758.1 | - | 6 - 7 | 2361 | 68 | |
M16801.1 | - | 7 - 8 | 2861 | 68 |