Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001044305.2 | NP_001037770.1 | 9 - 10 | 1218 | 78 |
NM_001281439.1 | NP_001268368.1 | 8 - 9 | 1137 | 78 | |
NM_001281440.1 | NP_001268369.1 | 9 - 10 | 1034 | 78 | |
NM_021940.4 | NP_068759.2 | 8 - 9 | 1137 | 78 | |
XM_011536045.2 | XP_011534347.1 | 9 - 10 | 1195 | 78 | |
XM_011536046.2 | XP_011534348.1 | 10 - 11 | 1120 | 78 | |
XM_011536047.2 | XP_011534349.1 | 8 - 9 | 1114 | 78 | |
XM_011536048.2 | XP_011534350.1 | 10 - 11 | 1107 | 78 | |
XM_017011170.1 | XP_016866659.1 | 10 - 11 | 1303 | 78 | |
GenBank mRNA | AF442495.1 | - | 8 - 9 | 1035 | 78 |
AK023221.1 | - | 8 - 9 | 952 | 78 | |
AK074340.1 | - | 1 - 2 | 52 | 78 | |
AK124239.1 | - | 10 - 11 | 1151 | 78 | |
AK222711.1 | - | 8 - 9 | 934 | 78 | |
AK290448.1 | - | 9 - 10 | 995 | 78 | |
AK307807.1 | - | 9 - 10 | 1031 | 78 | |
AK315579.1 | - | 8 - 9 | 1015 | 78 | |
AL833906.1 | - | 2 - 3 | 226 | 78 | |
AY055003.1 | - | 9 - 10 | 1033 | 78 | |
AY055004.1 | - | 8 - 9 | 952 | 78 | |
BC028074.1 | - | 8 - 9 | 1009 | 78 | |
BC036123.1 | - | 9 - 10 | 1071 | 78 |