Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014390.2 | NP_055205.2 | 3 - 4 | 574 | 68 |
XM_017011987.1 | XP_016867476.1 | 3 - 4 | 603 | 68 | |
GenBank mRNA | AB209510.1 | - | 3 - 4 | 512 | 68 |
AK096583.1 | - | 4 - 5 | 499 | 68 | |
AK312313.1 | - | 3 - 4 | 528 | 68 | |
BC017180.2 | - | 3 - 4 | 530 | 68 | |
BT009785.1 | - | 3 - 4 | 348 | 68 | |
U22055.1 | - | 3 - 4 | 540 | 68 |