Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014390.2 | NP_055205.2 | 8 - 9 | 1174 | 140 |
XM_017011987.1 | XP_016867476.1 | 8 - 9 | 1203 | 140 | |
GenBank mRNA | AK096583.1 | - | 9 - 10 | 1099 | 140 |
AK304181.1 | - | 8 - 9 | 1066 | 140 | |
AK312313.1 | - | 8 - 9 | 1128 | 140 | |
BC017180.2 | - | 8 - 9 | 1130 | 140 | |
BT009785.1 | - | 8 - 9 | 948 | 140 | |
U22055.1 | - | 8 - 9 | 1140 | 140 |