Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003922.3 | NP_003913.3 | 29 - 30 | 5629 | 79 |
XM_017022699.1 | XP_016878188.1 | 30 - 31 | 5735 | 79 | |
XM_017022700.1 | XP_016878189.1 | 30 - 31 | 5735 | 79 | |
XM_017022701.1 | XP_016878190.1 | 30 - 31 | 5735 | 79 | |
XM_017022702.1 | XP_016878191.1 | 29 - 30 | 5669 | 79 | |
XM_017022703.1 | XP_016878192.1 | 30 - 31 | 5735 | 79 | |
XM_017022704.1 | XP_016878193.1 | 30 - 31 | 5735 | 79 | |
XM_017022705.1 | XP_016878194.1 | 29 - 30 | 5642 | 79 | |
XM_017022706.1 | XP_016878195.1 | 30 - 31 | 5738 | 79 | |
GenBank mRNA | AK226162.1 | - | 16 - 17 | 2692 | 79 |
U50078.1 | - | 29 - 30 | 5577 | 79 |