Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003922.3 | NP_003913.3 | 64 - 65 | 12384 | 120 |
XM_017022699.1 | XP_016878188.1 | 65 - 66 | 12541 | 120 | |
XM_017022700.1 | XP_016878189.1 | 65 - 66 | 12505 | 120 | |
XM_017022701.1 | XP_016878190.1 | 65 - 66 | 12490 | 120 | |
XM_017022702.1 | XP_016878191.1 | 64 - 65 | 12490 | 120 | |
XM_017022703.1 | XP_016878192.1 | 65 - 66 | 12487 | 120 | |
XM_017022704.1 | XP_016878193.1 | 65 - 66 | 12469 | 120 | |
XM_017022705.1 | XP_016878194.1 | 64 - 65 | 12463 | 120 | |
GenBank mRNA | AK226162.1 | - | 51 - 52 | 9447 | 120 |
U50078.1 | - | 64 - 65 | 12332 | 120 |