Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003922.3 | NP_003913.3 | 75 - 76 | 14088 | 95 |
XM_017022699.1 | XP_016878188.1 | 76 - 77 | 14245 | 95 | |
XM_017022700.1 | XP_016878189.1 | 76 - 77 | 14209 | 95 | |
XM_017022701.1 | XP_016878190.1 | 76 - 77 | 14194 | 95 | |
XM_017022702.1 | XP_016878191.1 | 75 - 76 | 14194 | 95 | |
XM_017022703.1 | XP_016878192.1 | 76 - 77 | 14191 | 95 | |
XM_017022704.1 | XP_016878193.1 | 76 - 77 | 14173 | 95 | |
XM_017022705.1 | XP_016878194.1 | 75 - 76 | 14167 | 95 | |
GenBank mRNA | AK226162.1 | - | 62 - 63 | 11151 | 95 |
BC040929.1 | - | 9 - 10 | 1312 | 95 | |
U50078.1 | - | 75 - 76 | 14036 | 95 |