Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006456.2 | NP_006447.2 | 5 - 6 | 928 | 100 |
XM_005256954.3 | XP_005257011.1 | 5 - 6 | 931 | 100 | |
GenBank mRNA | AJ251053.1 | - | 5 - 6 | 712 | 100 |
AK225943.1 | - | 5 - 6 | 726 | 100 | |
BC040455.1 | - | 5 - 6 | 710 | 100 | |
BT019972.1 | - | 5 - 6 | 673 | 100 | |
U14550.1 | - | 5 - 6 | 712 | 100 |