Assay Design
Gene Transcripts
- Gene Symbol
- EXOG
- Entrez Gene ID
-
9941
- Gene Name
- endo/exonuclease (5'-3'), endonuclease G-like
- Gene Aliases
-
ENDOGL1, ENDOGL2, ENGL, ENGL-a, ENGL-b, ENGLA, ENGLB
- Chromosome Location
-
Chr.3: 38496127 - 38526305 on Build GRCh38
View Full Product Details