Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023008.3 | NP_075384.3 | 12 - 13 | 1221 | 112 |
XM_011528190.2 | XP_011526492.1 | 11 - 12 | 1152 | 112 | |
GenBank mRNA | AK023011.1 | - | 10 - 11 | 999 | 112 |
AK093879.1 | - | 11 - 12 | 1333 | 112 | |
AK097181.1 | - | 3 - 4 | 539 | 112 | |
AK300780.1 | - | 12 - 13 | 1205 | 112 | |
AK300850.1 | - | 12 - 13 | 1226 | 112 | |
AL833189.1 | - | 11 - 12 | 1874 | 112 | |
BC002890.2 | - | 9 - 10 | 886 | 112 | |
BC009876.2 | - | 3 - 4 | 393 | 112 | |
BC112249.1 | - | 9 - 10 | 878 | 112 | |
BC112251.1 | - | 9 - 10 | 878 | 112 | |
BC143770.1 | - | 9 - 10 | 878 | 112 |