Assay Design



Gene Transcripts

Gene Symbol
PLEKHB2
Entrez Gene ID
55041
Gene Name
pleckstrin homology domain containing B2
Gene Aliases
EVT2
Chromosome Location
Chr.2: 131104847 - 131149852 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100623.1 NP_001094093.1 4 - 5 851 104
NM_001267062.1 NP_001253991.1 4 - 5 851 104
NM_001267063.1 NP_001253992.1 4 - 5 851 104
NM_001267064.1 NP_001253993.1 4 - 5 851 104
NM_001267065.1 NP_001253994.1 4 - 5 851 104
NM_001267066.1 NP_001253995.1 4 - 5 851 104
NM_001309448.1 NP_001296377.1 3 - 4 806 104
NM_001309451.1 NP_001296380.1 3 - 4 806 104
NM_001309452.1 NP_001296381.1 5 - 6 949 104
NM_017958.2 NP_060428.2 4 - 5 851 104
XM_011511385.2 XP_011509687.1 4 - 5 390 104
XM_011511386.2 XP_011509688.1 4 - 5 393 104
XM_017004408.1 XP_016859897.1 3 - 4 319 104
XM_017004409.1 XP_016859898.1 3 - 4 319 104
XM_017004410.1 XP_016859899.1 3 - 4 319 104
XM_017004411.1 XP_016859900.1 5 - 6 1455 104
XM_017004412.1 XP_016859901.1 5 - 6 1455 104
GenBank mRNA AK000790.1 - 4 - 5 362 104
AK093730.1 - 4 - 5 1086 104
AK123710.1 - 4 - 5 808 104
AK126289.1 - 4 - 5 851 104
AK223338.1 - 4 - 5 362 104
AK293880.1 - 4 - 5 362 104
AK296008.1 - 3 - 4 315 104
BC001428.1 - 4 - 5 345 104
BC013991.2 - 4 - 5 344 104
BC050695.1 - 4 - 5 346 104
BP362174.1 - 3 - 4 317 104
BQ052811.1 - 4 - 5 343 104
BQ649140.1 - 4 - 5 360 104
BU174451.1 - 5 - 6 430 104
DB294071.1 - 4 - 5 364 104

View Full Product Details