Assay Design



Gene Transcripts

Gene Symbol
TICRR
Entrez Gene ID
90381
Gene Name
TOPBP1 interacting checkpoint and replication regulator
Gene Aliases
C15orf42, SLD3, Treslin
Chromosome Location
Chr.15: 89575587 - 89628023 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308025.1 NP_001294954.1 17 - 18 3018 79
NM_152259.3 NP_689472.3 17 - 18 3021 79
GenBank mRNA BX538335.1 - 4 - 5 322 79

View Full Product Details