Assay Design



Gene Transcripts

Gene Symbol
CCNE1
Entrez Gene ID
898
Gene Name
cyclin E1
Gene Aliases
CCNE, pCCNE1
Chromosome Location
Chr.19: 29811994 - 29824317 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011527440.2 XP_011525742.1 3 - 4 185 69

View Full Product Details