Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024086.3 | NP_076991.3 | 9 - 10 | 1211 | 99 |
XM_005256801.2 | XP_005256858.1 | 8 - 9 | 1048 | 99 | |
XM_017025045.1 | XP_016880534.1 | 10 - 11 | 1254 | 99 | |
XM_017025046.1 | XP_016880535.1 | 7 - 8 | 962 | 99 | |
GenBank mRNA | AK027410.1 | - | 9 - 10 | 1191 | 99 |
AK294547.1 | - | 9 - 10 | 1191 | 99 | |
AK296464.1 | - | 7 - 8 | 936 | 99 | |
AL832612.2 | - | 9 - 10 | 1170 | 99 | |
BC050603.1 | - | 9 - 10 | 1151 | 99 | |
BX647255.1 | - | 8 - 9 | 985 | 99 |