Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000189.4 | NP_000180.2 | 12 - 13 | 3766 | 61 |
XM_017003945.1 | XP_016859434.1 | 11 - 12 | 2150 | 61 | |
GenBank mRNA | AF148513.1 | - | 12 - 13 | 2289 | 61 |
AK290357.1 | - | 12 - 13 | 2269 | 61 | |
BC021116.1 | - | 12 - 13 | 2284 | 61 | |
BC064369.1 | - | 12 - 13 | 2282 | 61 | |
CR749202.1 | - | 12 - 13 | 1946 | 61 |