Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000189.4 | NP_000180.2 | 13 - 14 | 3863 | 59 |
XM_005264280.1 | XP_005264337.1 | 13 - 14 | 2406 | 59 | |
XM_011532807.1 | XP_011531109.1 | 12 - 13 | 2310 | 59 | |
XM_017003945.1 | XP_016859434.1 | 12 - 13 | 2247 | 59 | |
GenBank mRNA | AF148513.1 | - | 13 - 14 | 2386 | 59 |
AK290357.1 | - | 13 - 14 | 2366 | 59 | |
BC021116.1 | - | 13 - 14 | 2381 | 59 | |
BC064369.1 | - | 13 - 14 | 2379 | 59 |