Assay Design



Gene Transcripts

Gene Symbol
HK2
Entrez Gene ID
3099
Gene Name
hexokinase 2
Gene Aliases
HKII, HXK2
Chromosome Location
Chr.2: 74832655 - 74893354 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000189.4 NP_000180.2 15 - 16 4142 72
XM_005264280.1 XP_005264337.1 15 - 16 2685 72
XM_011532807.1 XP_011531109.1 14 - 15 2589 72
XM_017003945.1 XP_016859434.1 14 - 15 2526 72
GenBank mRNA AF148513.1 - 15 - 16 2665 72
AK290357.1 - 15 - 16 72
BC021116.1 - 15 - 16 2660 72
BC064369.1 - 15 - 16 2658 72
CR749202.1 - 15 - 16 72

View Full Product Details