Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000189.4 | NP_000180.2 | 4 - 5 | 2426 | 67 |
XM_005264280.1 | XP_005264337.1 | 4 - 5 | 906 | 67 | |
GenBank mRNA | AF148513.1 | - | 4 - 5 | 949 | 67 |
AK290357.1 | - | 4 - 5 | 929 | 67 | |
BC021116.1 | - | 4 - 5 | 944 | 67 | |
BC064369.1 | - | 4 - 5 | 942 | 67 | |
CR749202.1 | - | 4 - 5 | 606 | 67 |