Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257328.1 | NP_001244257.1 | 4 - 5 | 381 | 83 |
NM_001257329.1 | NP_001244258.1 | 4 - 5 | 381 | 83 | |
NM_001257330.1 | NP_001244259.1 | 4 - 5 | 381 | 83 | |
NM_004313.3 | NP_004304.1 | 4 - 5 | 381 | 83 | |
NR_047516.1 | 3 - 4 | 350 | 83 | ||
XM_011523858.2 | XP_011522160.1 | 4 - 5 | 266 | 83 | |
XM_017024645.1 | XP_016880134.1 | 3 - 4 | 253 | 83 | |
GenBank mRNA | AB209786.1 | - | 3 - 4 | 156 | 83 |
AF106941.1 | - | 4 - 5 | 206 | 83 | |
AK297059.1 | - | 3 - 4 | 214 | 83 | |
AK297181.1 | - | 4 - 5 | 182 | 83 | |
BC007427.2 | - | 4 - 5 | 215 | 83 | |
BC039066.1 | - | 3 - 4 | 144 | 83 | |
BC067368.1 | - | 4 - 5 | 209 | 83 | |
BC095450.1 | - | 4 - 5 | 179 | 83 | |
BG723452.1 | - | 4 - 5 | 391 | 83 | |
CR450310.1 | - | 4 - 5 | 153 | 83 | |
CR749218.1 | - | 3 - 4 | 174 | 83 | |
DQ664180.1 | - | 4 - 5 | 153 | 83 | |
EU883572.1 | - | 4 - 5 | 153 | 83 | |
Z11501.1 | - | 4 - 5 | 153 | 83 |