Assay Design



Gene Transcripts

Gene Symbol
ODF3B
Entrez Gene ID
440836
Gene Name
outer dense fiber of sperm tails 3B
Gene Aliases
ODF3L3
Chromosome Location
Chr.22: 50530409 - 50532579 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014440.3 NP_001014440.2 1 - 2 105 79
GenBank mRNA AW291097.1 - 2 - 3 79

View Full Product Details