Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014440.3 | NP_001014440.2 | 3 - 4 | 447 | 134 |
GenBank mRNA | BC127935.1 | - | 2 - 3 | 276 | 134 |
BG149596.1 | - | 1 - 2 | 180 | 134 | |
CD674272.1 | - | 2 - 3 | 346 | 134 |