Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256303.1 | NP_001243232.1 | 15 - 16 | 1818 | 77 |
NM_001256304.1 | NP_001243233.1 | 14 - 15 | 1555 | 77 | |
NM_001256308.2 | NP_001243237.1 | 14 - 15 | 1689 | 77 | |
NM_001320932.1 | NP_001307861.1 | 14 - 15 | 1663 | 77 | |
NM_001320933.1 | NP_001307862.1 | 14 - 15 | 1728 | 77 | |
NM_001320935.1 | NP_001307864.1 | 13 - 14 | 1577 | 77 | |
NM_183361.2 | NP_899205.1 | 14 - 15 | 1728 | 77 | |
XM_005264182.1 | XP_005264239.1 | 15 - 16 | 1735 | 77 | |
XM_011532673.1 | XP_011530975.1 | 15 - 16 | 1753 | 77 | |
XM_011532675.1 | XP_011530977.1 | 15 - 16 | 1753 | 77 | |
XM_017003529.1 | XP_016859018.1 | 14 - 15 | 1663 | 77 | |
XM_017003530.1 | XP_016859019.1 | 14 - 15 | 1645 | 77 | |
XM_017003531.1 | XP_016859020.1 | 14 - 15 | 1645 | 77 | |
XM_017003534.1 | XP_016859023.1 | 14 - 15 | 1663 | 77 | |
XM_017003535.1 | XP_016859024.1 | 14 - 15 | 1645 | 77 | |
XM_017003538.1 | XP_016859027.1 | 14 - 15 | 1581 | 77 | |
GenBank mRNA | AK225724.1 | - | 14 - 15 | 1566 | 77 |
AK294174.1 | - | 13 - 14 | 1482 | 77 | |
AK300019.1 | - | 15 - 16 | 1695 | 77 | |
AK301386.1 | - | 14 - 15 | 1691 | 77 | |
BC016655.1 | - | 14 - 15 | 1621 | 77 | |
BT009805.1 | - | 13 - 14 | 1465 | 77 | |
DQ160290.1 | - | 14 - 15 | 1555 | 77 | |
HY302507.1 | - | 5 - 6 | 77 |