Assay Design
Gene Transcripts
- Gene Symbol
- CLN3
- Entrez Gene ID
-
1201
- Gene Name
- CLN3, battenin
- Gene Aliases
-
BTS, JNCL
- Chromosome Location
-
Chr.16: 28466653 - 28492302 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF015595.1 |
- |
6 - 7 |
556 |
107 |
AF077960.1 |
- |
7 - 8 |
538 |
107 |
View Full Product Details