Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006788.3 | NP_006779.1 | 4 - 5 | 1267 | 124 |
GenBank mRNA | AB209924.1 | - | 2 - 3 | 474 | 124 |
AY517554.1 | - | 2 - 3 | 533 | 124 | |
BC013126.1 | - | 4 - 5 | 1129 | 124 | |
L42542.1 | - | 4 - 5 | 1275 | 124 |