Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000267.3 | NP_000258.1 | 25 - 26 | 3697 | 81 |
NM_001042492.2 | NP_001035957.1 | 25 - 26 | 3697 | 81 | |
XM_005257983.2 | XP_005258040.1 | 25 - 26 | 3647 | 81 | |
XM_005257984.2 | XP_005258041.1 | 25 - 26 | 3647 | 81 | |
XM_006721922.2 | XP_006721985.1 | 26 - 27 | 3677 | 81 | |
XM_006721924.2 | XP_006721987.1 | 26 - 27 | 3677 | 81 | |
XM_006721925.2 | XP_006721988.1 | 26 - 27 | 3677 | 81 | |
XM_006721926.3 | XP_006721989.1 | 26 - 27 | 3677 | 81 | |
XM_011524852.2 | XP_011523154.1 | 26 - 27 | 3674 | 81 | |
XM_011524857.2 | XP_011523159.1 | 26 - 27 | 3677 | 81 | |
XM_017024689.1 | XP_016880178.1 | 26 - 27 | 3677 | 81 | |
XM_017024690.1 | XP_016880179.1 | 26 - 27 | 3677 | 81 | |
GenBank mRNA | AB209336.1 | - | 8 - 9 | 1850 | 81 |
BC144643.1 | - | 25 - 26 | 3416 | 81 | |
BX537850.1 | - | 23 - 24 | 3089 | 81 | |
M38106.1 | - | 17 - 18 | 2312 | 81 | |
M38107.1 | - | 17 - 18 | 2312 | 81 | |
M82814.1 | - | 25 - 26 | 3525 | 81 | |
M89914.1 | - | 25 - 26 | 3511 | 81 |