Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000267.3 | NP_000258.1 | 2 - 3 | 588 | 196 |
NM_001042492.2 | NP_001035957.1 | 2 - 3 | 588 | 196 | |
NM_001128147.2 | NP_001121619.1 | 2 - 3 | 588 | 196 | |
XM_005257983.2 | XP_005258040.1 | 2 - 3 | 538 | 196 | |
XM_005257984.2 | XP_005258041.1 | 2 - 3 | 538 | 196 | |
XM_006721922.2 | XP_006721985.1 | 2 - 3 | 538 | 196 | |
XM_006721924.2 | XP_006721987.1 | 2 - 3 | 538 | 196 | |
XM_006721925.2 | XP_006721988.1 | 2 - 3 | 538 | 196 | |
XM_006721926.3 | XP_006721989.1 | 2 - 3 | 538 | 196 | |
XM_011524852.2 | XP_011523154.1 | 2 - 3 | 538 | 196 | |
XM_011524857.2 | XP_011523159.1 | 2 - 3 | 538 | 196 | |
XM_017024689.1 | XP_016880178.1 | 2 - 3 | 538 | 196 | |
XM_017024690.1 | XP_016880179.1 | 2 - 3 | 538 | 196 | |
GenBank mRNA | AK289936.1 | - | 2 - 3 | 538 | 196 |
BC144643.1 | - | 2 - 3 | 307 | 196 | |
D12625.1 | - | 2 - 3 | 588 | 196 | |
D42072.1 | - | 2 - 3 | 270 | 196 | |
M82814.1 | - | 2 - 3 | 416 | 196 | |
M89914.1 | - | 2 - 3 | 402 | 196 |