Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000267.3 | NP_000258.1 | 42 - 43 | 6957 | 69 |
NM_001042492.2 | NP_001035957.1 | 43 - 44 | 7020 | 69 | |
XM_005257983.2 | XP_005258040.1 | 43 - 44 | 6970 | 69 | |
XM_005257984.2 | XP_005258041.1 | 42 - 43 | 6907 | 69 | |
XM_006721922.2 | XP_006721985.1 | 44 - 45 | 7000 | 69 | |
XM_006721924.2 | XP_006721987.1 | 44 - 45 | 7000 | 69 | |
XM_006721925.2 | XP_006721988.1 | 43 - 44 | 6937 | 69 | |
XM_006721926.3 | XP_006721989.1 | 44 - 45 | 7000 | 69 | |
XM_011524852.2 | XP_011523154.1 | 44 - 45 | 6997 | 69 | |
XM_011524857.2 | XP_011523159.1 | 44 - 45 | 7000 | 69 | |
XM_017024689.1 | XP_016880178.1 | 43 - 44 | 6937 | 69 | |
GenBank mRNA | BC144643.1 | - | 43 - 44 | 6773 | 69 |
M38106.1 | - | 34 - 35 | 5575 | 69 | |
M38107.1 | - | 34 - 35 | 5575 | 69 | |
M82814.1 | - | 42 - 43 | 6785 | 69 | |
M89914.1 | - | 43 - 44 | 6834 | 69 |